Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020309.3 | NP_064705.1 | 3 | 573 | ||
GenBank mRNA | AB032436.1 | - | 3 | 422 | ||
AK057879.1 | - | 519 | ||||
AK095659.2 | - | 2 | 1852 | |||
AK294226.1 | - | 3 | 379 | |||
AK294405.1 | - | 3 | 422 | |||
BC059379.1 | - | 3 | 548 | 5262415 | ||
BG701511.1 | - | 3 | 549 |