Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021063.3 | NP_066407.1 | 1 | 165 | ||
NM_138720.2 | NP_619790.1 | 1 | 165 | |||
XM_005249039.3 | XP_005249096.1 | 1 | 301 | |||
GenBank mRNA | AJ223353.1 | - | 1 | 144 | ||
AK292712.1 | - | 1 | 161 | |||
BC002842.2 | - | 1 | 141 | 3659807 | ||
BC096122.3 | - | 1 | 116 | 40002354 | ||
BE245851.1 | - | 1 | 165 | |||
BM153714.1 | - | 1 | 165 |