Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286655.1 | NP_001273584.1 | 6 | 1342 | ||
NM_001286656.1 | NP_001273585.1 | 6 | 1019 | |||
NM_023932.3 | NP_076421.2 | 6 | 1037 | |||
NM_206539.2 | NP_996262.1 | 6 | 1453 | |||
XM_005249308.4 | XP_005249365.1 | 5 | 2245 | |||
XM_011514823.2 | XP_011513125.1 | 5 | 1606 | |||
XM_017011204.1 | XP_016866693.1 | 3 | 1055 | |||
GenBank mRNA | AK055380.1 | - | 6 | 1453 | ||
AY358126.1 | - | 6 | 903 | |||
BC000230.1 | - | 6 | 736 | 3350499 | ||
BC006425.1 | - | 6 | 736 | 3349698 | ||
BC110320.1 | - | 6 | 1005 | 5495458 |