Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303451.1 | NP_001290380.1 | 3 | 366 | ||
NM_024731.3 | NP_079007.2 | 3 | 366 | |||
XM_005256149.2 | XP_005256206.1 | 3 | 580 | |||
XM_017023674.1 | XP_016879163.1 | 3 | 328 | |||
XM_017023675.1 | XP_016879164.1 | 3 | 348 | |||
GenBank mRNA | AK022605.1 | - | 2 | 219 | ||
BC032400.1 | - | 3 | 342 | 5210986 | ||
BC033821.1 | - | 3 | 352 | 5219021 | ||
DA049618.1 | - | 3 | 368 |