Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003374.2 | NP_003365.1 | 7 | 890 | ||
NR_036624.1 | 7 | 968 | ||||
NR_036625.1 | 7 | 971 | ||||
XM_005272075.3 | XP_005272132.1 | 7 | 887 | |||
XM_017009821.1 | XP_016865310.1 | 7 | 818 | |||
XM_017009822.1 | XP_016865311.1 | 7 | 797 | |||
XM_017009823.1 | XP_016865312.1 | 7 | 820 | |||
GenBank mRNA | AK095989.1 | - | 7 | 890 | ||
AK122953.1 | - | 10 | 2293 | |||
BC008482.1 | - | 7 | 676 | 4332199 | ||
BC071168.1 | - | 7 | 707 | 6023095 | ||
BC090042.1 | - | 7 | 700 | 30395350 | ||
L06132.1 | - | 7 | 744 |