Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165881.2 | NP_001159353.1 | 3 | 302 | ||
NM_001165882.2 | NP_001159354.1 | 3 | 309 | |||
NM_001165883.1 | NP_001159355.1 | 3 | 426 | |||
NM_001165885.1 | NP_001159357.1 | 3 | 426 | |||
NM_003415.2 | NP_003406.1 | 3 | 426 | |||
NM_152943.2 | NP_694422.2 | 3 | 426 | |||
GenBank mRNA | AF141655.1 | - | 3 | 276 | ||
AF317549.1 | - | 3 | 426 | |||
AF385187.1 | - | 3 | 276 | |||
AK027712.1 | - | 3 | 340 | |||
AY583691.1 | - | 3 | 257 | |||
BC110542.2 | - | 3 | 302 | 40036577 | ||
CN255799.1 | - | 3 | 286 | |||
DQ057356.1 | - | 3 | 215 | |||
DQ057358.1 | - | 3 | 215 |