Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030978.2 | NP_112240.1 | 4, 5 | 1479 | ||
XM_011519067.1 | XP_011517369.1 | 2, 3 | 613 | |||
GenBank mRNA | AF087842.1 | - | 2, 3 | 267 | ||
AK056174.1 | - | 4, 5 | 1467 | |||
BC000018.1 | - | 2, 3 | 269 | 3509794 | ||
BC000798.1 | - | 2, 3 | 301 | 3451934 | ||
BC002418.1 | - | 2, 3 | 269 | 3163950 | ||
BX537659.1 | - | 2, 3 | 3748 |