Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145165.1 | NP_001138637.1 | 3 | 767 | ||
NM_031304.4 | NP_112594.1 | 3 | 478 | |||
XM_011528341.2 | XP_011526643.1 | 3 | 475 | |||
GenBank mRNA | BC002817.2 | - | 3 | 478 | 3635058 | |
BC009863.2 | - | 2 | 367 | 3938535 |