Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270440.1 | NP_001257369.1 | 2, 3 | 416 | ||
NM_031429.2 | NP_113617.1 | 2, 3 | 416 | |||
XM_005260089.1 | XP_005260146.1 | 2, 3 | 416 | |||
XM_011528343.1 | XP_011526645.1 | 1, 2 | 82 | |||
XM_017027343.1 | XP_016882832.1 | 2, 3 | 416 | |||
XM_017027344.1 | XP_016882833.1 | 2, 3 | 416 | |||
GenBank mRNA | AK093672.1 | - | 2, 3 | 416 | ||
BC005063.1 | - | 2, 3 | 404 | 3344818 | ||
DB106146.1 | - | 2, 3 | 416 |