Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF212239.1 | - | 1, 2 | 174 | ||
AK223378.1 | - | 1, 2 | 189 | |||
AK311977.1 | - | 1, 2 | 187 | |||
AY007436.1 | - | 1, 2 | 170 | |||
BC029355.1 | - | 1, 2 | 191 | 4613026 |