Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032377.3 | NP_115753.1 | 2 | 168 | ||
XM_017027356.1 | XP_016882845.1 | 3 | 323 | |||
XM_017027357.1 | XP_016882846.1 | 2 | 182 | |||
XM_017027358.1 | XP_016882847.1 | 2 | 322 | |||
XM_017027359.1 | XP_016882848.1 | 2 | 238 | |||
GenBank mRNA | AK001171.1 | - | 3 | 325 | ||
AK097850.1 | - | 2 | 162 | |||
BC007516.2 | - | 1 | 123 | 3028469 | ||
BG718042.1 | - | 2 | 272 |