Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009998.3 | NP_001009998.1 | 2, 3 | 438 | ||
NM_032627.4 | NP_116016.1 | 2, 3 | 438 | |||
XM_005259790.3 | XP_005259847.1 | 2, 3 | 479 | |||
XM_006722665.3 | XP_006722728.1 | 2, 3 | 485 | |||
XM_006722666.2 | XP_006722729.1 | 2, 3 | 443 | |||
XM_006722668.2 | XP_006722731.1 | 2, 3 | 443 | |||
XM_017026431.1 | XP_016881920.1 | 2, 3 | 497 | |||
XM_017026432.1 | XP_016881921.1 | 2, 3 | 1035 | |||
XM_017026433.1 | XP_016881922.1 | 2, 3 | 443 | |||
XM_017026434.1 | XP_016881923.1 | 2, 3 | 492 | |||
XM_017026435.1 | XP_016881924.1 | 2, 3 | 1034 | |||
XM_017026436.1 | XP_016881925.1 | 2, 3 | 443 | |||
XM_017026437.1 | XP_016881926.1 | 2, 3 | 474 | |||
GenBank mRNA | AK098429.1 | - | 2, 3 | 344 | ||
BC000274.1 | - | 2, 3 | 345 | 3356283 | ||
BF207000.1 | - | 2, 3 | 379 | |||
BQ068310.1 | - | 477 | ||||
BU838485.1 | - | 2 | 209 | |||
CX873722.1 | - | 2, 3 | 438 |