Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080671.3 | NP_542402.3 | 2 | 481 | ||
GenBank mRNA | AK294423.1 | - | 2 | 481 | ||
AK313203.1 | - | 2 | 221 | |||
AY065987.1 | - | 2 | 195 | |||
BC014429.1 | - | 2 | 202 | 4548085 | ||
BP348485.1 | - | 2 | 267 | |||
BP359441.1 | - | 2 | 221 | |||
BP362695.1 | - | 2 | 221 | |||
DQ192293.1 | - | 2 | 222 |