Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286135.1 | NP_001273064.1 | 2 | 397 | ||
NM_031492.3 | NP_113680.1 | 2 | 397 | |||
XM_011545297.2 | XP_011543599.1 | 2 | 363 | |||
XM_017018402.1 | XP_016873891.1 | 2 | 363 | |||
GenBank mRNA | AK095158.1 | - | 1 | 896 | ||
BC003503.1 | - | 2 | 339 | 3345085 | ||
BC004951.1 | - | 2 | 280 | 3620125 | ||
BQ428264.1 | - | 2 | 388 | |||
DB465480.1 | - | 2 | 397 |