Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032488.3 | NP_115877.2 | 4 | 397 | ||
XM_005259332.2 | XP_005259389.1 | 5 | 502 | |||
XM_011527396.1 | XP_011525698.1 | 5 | 497 | |||
XM_011527397.1 | XP_011525699.1 | 5 | 443 | |||
GenBank mRNA | AB049591.2 | - | 4 | 392 | ||
BC101197.1 | - | 5 | 453 | 40021735 | ||
BC101198.2 | - | 4 | 373 | 40021738 |