Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 12 | 2670 | ||
GenBank mRNA | AF245044.1 | - | 1 | 989 | ||
BC056664.1 | - | 1 | 695 |