Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164269.1 | NP_001157741.1 | 9 | 959 | ||
NM_002875.4 | NP_002866.2 | 9 | 1111 | |||
NM_133487.3 | NP_597994.3 | 9 | 1114 | |||
XM_006720626.3 | XP_006720689.1 | 9 | 1162 | |||
XM_011521857.2 | XP_011520159.1 | 9 | 861 | |||
XM_011521858.2 | XP_011520160.1 | 9 | 908 | |||
XM_011521859.2 | XP_011520161.1 | 9 | 1045 | |||
XM_011521860.2 | XP_011520162.1 | 9 | 956 | |||
XM_011521862.2 | XP_011520164.1 | 8 | 961 | |||
GenBank mRNA | AK131299.1 | - | 9 | 959 | ||
AK291969.1 | - | 9 | 1069 | |||
AK313503.1 | - | 9 | 1066 | |||
BC001459.2 | - | 7 | 777 | 3139011 | ||
BT019705.1 | - | 8 | 812 | |||
CR536559.1 | - | 8 | 812 | |||
D13804.1 | - | 9 | 1023 | |||
D14134.1 | - | 9 | 1044 |