Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133494.2 | NP_598001.1 | 3 | 507 | ||
XM_011509209.1 | XP_011507511.1 | 3 | 485 | |||
XM_017000344.1 | XP_016855833.1 | 3 | 485 | |||
XM_017000345.1 | XP_016855834.1 | 4 | 592 | |||
XM_017000346.1 | XP_016855835.1 | 3 | 485 | |||
XM_017000347.1 | XP_016855836.1 | 3 | 485 | |||
XM_017000348.1 | XP_016855837.1 | 3 | 485 | |||
GenBank mRNA | AB062450.1 | - | 2 | 226 | ||
AK313411.1 | - | 3 | 307 | |||
DB498485.1 | - | 3 | 507 |