Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160407.1 | NP_001153879.1 | 2 | 194 | ||
NM_138463.3 | NP_612472.1 | 2 | 327 | |||
XM_006721671.3 | XP_006721734.1 | 2 | 130 | |||
XM_011524278.2 | XP_011522580.1 | 2 | 1770 | |||
GenBank mRNA | BC014072.2 | - | 2 | 323 | 3957355 | |
CA453962.1 | - | 2 | 333 |