Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145400.1 | NP_001138872.1 | 3, 4 | 724 | ||
NM_139174.3 | NP_631913.3 | 4, 5 | 940 | |||
XM_005255814.1 | XP_005255871.1 | 3, 4 | 724 | |||
XM_011522919.1 | XP_011521221.1 | 4, 5 | 940 | |||
XM_011522920.1 | XP_011521222.1 | 4, 5 | 635 | |||
GenBank mRNA | AF447586.1 | - | 3, 4 | 662 | ||
AK093046.1 | - | 4, 5 | 919 | |||
AK093523.1 | - | 1, 2 | 1260 | |||
AK131102.1 | - | 4, 5 | 3488 | |||
AK315171.1 | - | 3, 4 | 643 | |||
BC033491.1 | - | 3, 4 | 662 | 5167849 |