Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185009.2 | NP_001171938.1 | 7 | 1183 | ||
NM_002938.4 | NP_002929.1 | 6 | 661 | |||
GenBank mRNA | AB000468.1 | - | 6 | 610 | ||
AK128038.1 | - | 6 | 1824 | |||
AK312534.1 | - | 6 | 590 | |||
U95140.1 | - | 6 | 583 |