Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277074.1 | NP_001264003.1 | 2 | 157 | ||
NM_144716.5 | NP_653317.2 | 3 | 312 | |||
NR_102269.1 | 2 | 175 | ||||
XM_011533390.2 | XP_011531692.1 | 2 | 252 | |||
XM_011533391.2 | XP_011531693.1 | 3 | 318 | |||
XM_011533392.2 | XP_011531694.1 | 3 | 270 | |||
XM_011533394.2 | XP_011531696.1 | 2 | 794 | |||
XM_011533396.2 | XP_011531698.1 | 2 | 511 | |||
GenBank mRNA | AK096749.1 | - | 2 | 135 | ||
AK295532.1 | - | 2 | 135 | |||
AK303160.1 | - | 2 | 149 | |||
BC020830.1 | - | 2 | 138 | 4771328 | ||
BM803652.1 | - | 2 | 82 | |||
CN430023.1 | - | 3 | 267 | |||
CV574708.1 | - | 4 | 388 | |||
DA952452.1 | - | 175 | ||||
DC379144.1 | - | 2 | 160 |