Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287005.1 | NP_001273934.1 | 3 | 298 | ||
NM_001287006.1 | NP_001273935.1 | 2 | 184 | |||
NM_145006.3 | NP_659443.1 | 2 | 184 | |||
XM_011518358.1 | XP_011516660.1 | 5 | 1378 | |||
XM_011518359.1 | XP_011516661.1 | 4 | 1328 | |||
XM_011518361.2 | XP_011516663.1 | 5 | 440 | |||
XM_011518362.1 | XP_011516664.1 | 3 | 1214 | |||
XM_017014448.1 | XP_016869937.1 | 3 | 485 | |||
XM_017014449.1 | XP_016869938.1 | 2 | 366 | |||
XM_017014450.1 | XP_016869939.1 | 4 | 325 | |||
XM_017014451.1 | XP_016869940.1 | 2 | 934 | |||
GenBank mRNA | AK128289.1 | - | 1 | 279 | ||
AY358190.1 | - | 3 | 226 | |||
BC014601.2 | - | 2 | 165 | 4821517 | ||
BC041834.1 | - | 1 | 205 | 5268318 | ||
DR007158.1 | - | 2 | 160 | |||
DT220823.1 | - | 2 | 184 |