Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145288.1 | NP_660331.1 | 1 | 129 | ||
GenBank mRNA | AF447583.1 | - | 2 | 175 | ||
BC019352.1 | - | 1 | 129 | 3942704 |