Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145886.3 | NP_665893.2 | 3 | 764 | ||
NM_145887.3 | NP_665894.2 | 3 | 764 | |||
XM_005253005.4 | XP_005253062.1 | 4 | 1702 | |||
XM_005253006.4 | XP_005253063.1 | 4 | 1265 | |||
XM_005253007.3 | XP_005253064.1 | 3 | 782 | |||
XM_005253008.4 | XP_005253065.1 | 4 | 1762 | |||
XM_011520209.2 | XP_011518511.1 | 3 | 1000 | |||
XM_011520210.2 | XP_011518512.1 | 4 | 1702 | |||
XM_011520211.2 | XP_011518513.1 | 4 | 1702 | |||
XM_011520212.1 | XP_011518514.1 | 3 | 842 | |||
XM_011520213.2 | XP_011518515.1 | 1 | 21 | |||
GenBank mRNA | AB208832.1 | - | 2 | 1286 | ||
AB208949.1 | - | 2 | 1250 | |||
AB209529.1 | - | 2 | 1392 | |||
AF229178.1 | - | 2 | 966 | |||
AF274972.1 | - | 3 | 760 | |||
BC014904.1 | - | 2 | 702 | 3908102 | ||
BQ059356.1 | - | 3 | 752 | |||
BX349115.2 | - | 1 | 117 |