Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007137.3 | NP_009068.2 | 5 | 1616 | ||
XM_005272600.3 | XP_005272657.1 | 5 | 1674 | |||
XM_011543899.2 | XP_011542201.1 | 5 | 3280 | |||
XM_011543900.2 | XP_011542202.1 | 6 | 1735 | |||
XM_017029486.1 | XP_016884975.1 | 6 | 1793 | |||
XM_017029487.1 | XP_016884976.1 | 5 | 2026 | |||
GenBank mRNA | AY487248.1 | - | 4 | 1367 |
Gene Symbol: ZNF585A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288800.1 | NP_001275729.1 | 5 | 1530 | ||
NM_152655.3 | NP_689868.1 | 6 | 1632 | |||
NM_199126.2 | NP_954577.1 | 6 | 1572 | |||
GenBank mRNA | AK074345.1 | - | 3 | 1164 | ||
AK290660.1 | - | 5 | 1509 | |||
BC052956.1 | - | 6 | 1572 | |||
BC063820.1 | - | 5 | 1464 | 6500797 |