Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152901.3 | NP_690865.1 | 1 | 211 | ||
GenBank mRNA | AF086332.1 | - | 1 | 211 | ||
AF454669.1 | - | 1 | 200 | |||
AF467809.1 | - | 1 | 206 | |||
AK313418.1 | - | 1 | 477 | |||
AY163727.1 | - | 1 | 211 | |||
BC105033.1 | - | 1 | 197 | 8144036 | ||
BC105035.1 | - | 1 | 197 | 8144038 |