Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258211.1 | NP_001245140.1 | 6, 7 | 953 | ||
NM_172366.3 | NP_758954.1 | 7, 8 | 989 | |||
GenBank mRNA | AF453435.1 | - | 7, 8 | 898 | ||
BC074986.2 | - | 7, 8 | 888 | 30915303 | ||
BC102026.1 | - | 7, 8 | 908 | 40031445 | ||
BC102027.1 | - | 6, 7 | 872 | 40031444 | ||
BC102028.1 | - | 6, 7 | 872 | 40031440 |