Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144661.2 | NP_653262.1 | 4, 5 | 525 | ||
XM_011539347.2 | XP_011537649.1 | 4, 5 | 962 | |||
GenBank mRNA | BC021737.2 | - | 4, 5 | 525 | 4821913 |