Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145270.2 | NP_660313.1 | 2 | 431 | ||
XM_017022959.1 | XP_016878448.1 | 2 | 641 | |||
XM_017022960.1 | XP_016878449.1 | 3 | 1164 | |||
XM_017022961.1 | XP_016878450.1 | 2 | 376 | |||
GenBank mRNA | BC016613.1 | - | 2 | 431 | 3874510 |