Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318878.1 | NP_001305807.1 | 5 | 840 | ||
NM_001318879.1 | NP_001305808.1 | 5 | 701 | |||
NM_173575.3 | NP_775846.2 | 5 | 802 | |||
NR_134911.1 | 5 | 479 | ||||
XM_011539688.1 | XP_011537990.1 | 5 | 873 | |||
XM_011539690.2 | XP_011537992.1 | 5 | 788 | |||
XM_011539693.2 | XP_011537995.1 | 5 | 806 | |||
XM_011539694.1 | XP_011537996.1 | 5 | 540 | |||
XM_011539695.1 | XP_011537997.1 | 5 | 741 | |||
XM_011539696.1 | XP_011537998.1 | 5 | 548 | |||
GenBank mRNA | AK057849.1 | - | 5 | 479 | ||
AK098366.1 | - | 4 | 639 | |||
AK302102.1 | - | 5 | 787 | |||
AK308956.1 | - | 5 | 879 | |||
AK310725.1 | - | 5 | 806 | |||
AY098866.1 | - | 5 | 802 | |||
BC045760.1 | - | 5 | 691 | 4831007 |