Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 2008 | ||
NM_001256896.1 | NP_001243825.1 | 4 | 1717 | |||
NM_173660.4 | NP_775931.3 | 7 | 2019 | |||
GenBank mRNA | AK091037.1 | - | 7 | 2003 | ||
AK096456.1 | - | 4 | 1717 | |||
BC043568.1 | - | 3 | 1356 | |||
BC062369.1 | - | 3 | 1411 | |||
BC131544.1 | - | 7 | 1956 | 40085498 | ||
BC141852.1 | - | 7 | 1990 | 8860352 |