Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318025.1 | NP_001304954.1 | 8 | 1202 | ||
NM_001318026.1 | NP_001304955.1 | 9 | 1144 | |||
NM_001318027.1 | NP_001304956.1 | 8 | 1140 | |||
NM_174907.3 | NP_777567.1 | 9 | 1373 | |||
GenBank mRNA | AF327345.1 | - | 9 | 1190 | ||
AJ271448.2 | - | 2 | 1771 | |||
AK289901.1 | - | 9 | 1340 | |||
AL833216.1 | - | 6 | 789 | |||
BC110889.1 | - | 8 | 1159 | 6503859 | ||
BC128136.1 | - | 9 | 1149 | 40112443 | ||
BC128137.1 | - | 9 | 1149 | 40112444 |