Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257359.1 | NP_001244288.1 | 5 | 944 | ||
NM_174920.3 | NP_777580.1 | 5 | 944 | |||
XM_011524490.2 | XP_011522792.1 | 2 | 130 | |||
XM_017024322.1 | XP_016879811.1 | 4 | 1328 | |||
XM_017024323.1 | XP_016879812.1 | 6 | 1111 | |||
XM_017024324.1 | XP_016879813.1 | 5 | 1799 | |||
GenBank mRNA | AK094209.1 | - | 5 | 875 | ||
AK095809.1 | - | 2 | 245 | |||
AK225767.1 | - | 5 | 875 | |||
AK291482.1 | - | 5 | 871 | |||
AY148482.1 | - | 5 | 865 | |||
BC029872.1 | - | 1 | 56 | |||
BC047081.1 | - | 4 | 598 | |||
BC136485.1 | - | 5 | 857 | 9020474 | ||
BC141822.1 | - | 5 | 884 | 40148162 | ||
BC142728.1 | - | 5 | 841 | 40148957 |