Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198670.1 | NP_001185599.1 | 2 | 701 | ||
NM_001198671.1 | NP_001185600.1 | 2 | 382 | |||
NM_001198672.1 | NP_001185601.1 | 2 | 371 | |||
NM_001198673.1 | NP_001185602.1 | 2 | 357 | |||
NM_001198674.1 | NP_001185603.1 | 2 | 382 | |||
NM_174926.2 | NP_777586.1 | 2 | 701 | |||
XM_017017327.1 | XP_016872816.1 | 1 | 3285 | |||
GenBank mRNA | AK128040.1 | - | 2 | 386 | ||
AK289680.1 | - | 2 | 199 | |||
AK294713.1 | - | 2 | 210 | |||
AK304086.1 | - | 2 | 534 | |||
BC015232.1 | - | 2 | 523 | 3900115 | ||
DA097438.1 | - | 2 | 308 | |||
DA331965.1 | - | 2 | 187 |