Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177967.3 | NP_808882.1 | 1 | 589 | ||
NR_026644.1 | 1 | 589 | ||||
XM_006719948.3 | XP_006720011.1 | 2 | 155 | |||
XM_011521083.2 | XP_011519385.1 | 2 | 152 | |||
GenBank mRNA | AK054563.1 | - | 1 | 590 | ||
BC121138.1 | - | 1 | 188 | |||
BC121139.1 | - | 1 | 188 | 40120987 |