Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 7 | 699 | ||
NM_001258332.1 | NP_001245261.1 | 5 | 574 | |||
GenBank mRNA | AK096026.1 | - | 4 | 358 | ||
AK096137.1 | - | 6 | 738 | |||
AK293212.1 | - | 5 | 857 | |||
AK300070.1 | - | 7 | 588 | |||
AK303279.1 | - | 5 | 527 | |||
AK310049.1 | - | 4 | 932 | |||
AK314340.1 | - | 7 | 681 | |||
BC015045.2 | - | 7 | 624 | 3922902 | ||
BI910081.1 | - | 7 | 601 | |||
BT009852.1 | - | 7 | 583 | |||
KU178089.1 | - | 7 | 583 | |||
KU178090.1 | - | 7 | 650 | |||
M60091.1 | - | 7 | 611 |