Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000413.2 | NP_000404.2 | 1 | 1052 | ||
XM_005257292.3 | XP_005257349.1 | 1 | 719 | |||
XM_006721857.3 | XP_006721920.1 | 1 | 719 | |||
XM_006721858.3 | XP_006721921.1 | 1 | 719 | |||
XM_006721859.3 | XP_006721922.1 | 1 | 719 | |||
XM_011524729.1 | XP_011523031.1 | 1 | 460 | |||
XM_011524730.1 | XP_011523032.1 | 1 | 460 | |||
XM_011524731.1 | XP_011523033.1 | 1 | 460 | |||
XM_011524732.2 | XP_011523034.1 | 1 | 719 | |||
GenBank mRNA | AK127832.1 | - | 1 | 3804 | ||
AK300143.1 | - | 1 | 95 | |||
AK300159.1 | - | 1 | 97 | |||
AK300452.1 | - | 1 | 95 | |||
AK316292.1 | - | 1 | 280 | |||
BC104752.1 | - | 1 | 110 | 8143755 | ||
BC111935.1 | - | 1 | 110 | 8327403 | ||
DQ469954.1 | - | 1 | 84 | |||
M36263.1 | - | 1 | 93 | |||
X13440.1 | - | 1 | 93 |