Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000606.2 | NP_000597.2 | 1 | 194 | ||
GenBank mRNA | BC113624.1 | - | 1 | 169 | 8322676 | |
BC113626.1 | - | 1 | 169 | 8322678 | ||
BM852158.1 | - | 1 | 194 | |||
M17263.1 | - | 1 | 145 | |||
M17999.1 | - | 1 | 180 | |||
X06465.1 | - | 1 | 180 |