Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 3, 4 | 536 | ||
NM_003151.3 | NP_003142.1 | 3, 4 | 587 | |||
XM_006712719.3 | XP_006712782.1 | 3, 4 | 582 | |||
XM_011511705.2 | XP_011510007.1 | 6, 7 | 733 | |||
XM_017004784.1 | XP_016860273.1 | 3, 4 | 341 | |||
GenBank mRNA | AK299976.1 | - | 3, 4 | 308 | ||
AK300868.1 | - | 3, 4 | 340 | |||
BC031212.1 | - | 3, 4 | 508 | 4830583 | ||
L78440.1 | - | 3, 4 | 353 |