Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011.3 | NP_001002.1 | 3 | 207 | ||
GenBank mRNA | AB209386.1 | - | 2 | 353 | ||
AF085824.1 | - | 2 | 78 | |||
AK311794.1 | - | 3 | 145 | |||
BC002866.2 | - | 3 | 163 | 3940495 | ||
BC061901.1 | - | 3 | 200 | 4442840 | ||
BC071919.1 | - | 1 | 127 | 5811776 | ||
BG615667.1 | - | 3 | 208 | |||
M77233.1 | - | 2 | 96 |
Gene Symbol: LOC100287685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298720.1 | - | 1 | 1317 |