Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001344.3 | NP_001335.1 | 1 | 211 | ||
GenBank mRNA | AK223129.1 | - | 1 | 198 | ||
BC007403.2 | - | 1 | 183 | 2988797 | ||
BC009798.1 | - | 1 | 185 | 4284779 | ||
BP198316.1 | - | 1 | 211 | |||
CR407682.1 | - | 1 | 131 | |||
CR542204.1 | - | 1 | 131 | |||
D15057.1 | - | 1 | 197 |