Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142797.1 | NP_001136269.1 | 2 | 391 | ||
NM_001504.1 | NP_001495.1 | 2 | 147 | |||
XM_005262256.3 | XP_005262313.1 | 2 | 248 | |||
XM_005262257.3 | XP_005262314.1 | 2 | 248 | |||
XM_017029435.1 | XP_016884924.1 | 2 | 329 | |||
XM_017029436.1 | XP_016884925.1 | 2 | 145 | |||
GenBank mRNA | AF469635.1 | - | 2 | 384 | ||
AK313679.1 | - | 2 | 141 | |||
AY242128.1 | - | 1 | 79 | |||
BC034403.1 | - | 2 | 117 | 5176136 | ||
KU178101.1 | - | 1 | 79 | |||
KU178102.1 | - | 1 | 79 | |||
X95876.1 | - | 2 | 147 |