Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001662.3 | NP_001653.1 | 2 | 245 | ||
GenBank mRNA | AF493884.1 | - | 2 | 98 | ||
AK297026.1 | - | 2 | 150 | |||
AK307297.1 | - | 2 | 219 | |||
AK308548.1 | - | 2 | 427 | |||
AK312107.1 | - | 2 | 150 | |||
BC003043.1 | - | 2 | 135 | 2822973 | ||
BC033104.1 | - | 2 | 192 | 4862014 | ||
BT007087.1 | - | 2 | 98 | |||
CN259625.1 | - | 2 | 262 | |||
M57567.1 | - | 2 | 152 |