Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001803.2 | NP_001794.2 | 2 | 288 | ||
GenBank mRNA | A23013.1 | - | 2 | 223 | ||
BC000644.2 | - | 2 | 232 | 3345325 | ||
BI835519.1 | - | 2 | 290 | |||
FJ460513.1 | - | 2 | 288 | |||
X62466.1 | - | 2 | 223 | |||
X67699.1 | - | 2 | 214 |