Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001818.3 | NP_001809.3 | 2 | 139 | ||
GenBank mRNA | AB031085.1 | - | 2 | 106 | ||
AB045829.1 | - | 2 | 106 | |||
AK314988.1 | - | 2 | 180 | |||
BC020744.1 | - | 2 | 139 | 4734943 | ||
D26125.1 | - | 2 | 102 | |||
M33375.1 | - | 2 | 104 | |||
S68287.1 | - | 2 | 113 |