Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039847.2 | NP_001034936.1 | 2 | 255 | ||
NM_001039848.2 | NP_001034937.1 | 2 | 333 | |||
NM_002085.4 | NP_002076.2 | 2 | 255 | |||
GenBank mRNA | AK225859.1 | - | 2 | 174 | ||
AL558552.3 | - | 2 | 108 | |||
BC010157.2 | - | 2 | 83 | |||
BC011836.2 | - | 2 | 176 | 3945177 | ||
BC021567.1 | - | 2 | 169 | 4581069 | ||
BC022071.1 | - | 2 | 192 | 4821595 | ||
BC032695.1 | - | 2 | 176 | 5227887 | ||
BC039849.1 | - | 2 | 244 | 5744771 | ||
BC046163.1 | - | 2 | 151 | 5742889 | ||
BC058329.1 | - | 2 | 172 | |||
BI561548.1 | - | 2 | 251 | |||
BM764133.1 | - | 2 | 334 | |||
BQ648717.1 | - | 2 | 120 | |||
CF127510.1 | - | 2 | 267 | |||
HY049693.1 | - | 2 | 256 | |||
X71973.1 | - | 2 | 217 |