Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002281.3 | NP_002272.2 | 5 | 889 | ||
GenBank mRNA | BC006452.1 | - | 1 | 57 | ||
BC117465.1 | - | 6 | 860 | 40126016 | ||
BC117467.1 | - | 6 | 861 | 40126018 | ||
X80197.1 | - | 1 | 97 | |||
X81420.1 | - | 6 | 601 |