Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002452.3 | NP_002443.3 | 2 | 107 | ||
NM_198948.1 | NP_945186.1 | 3 | 158 | |||
NM_198949.1 | NP_945187.1 | 3 | 231 | |||
NM_198950.1 | NP_945188.1 | 3 | 155 | |||
NM_198952.1 | NP_945190.1 | 3 | 228 | |||
NM_198953.1 | NP_945191.1 | 2 | 143 | |||
NM_198954.1 | NP_945192.1 | 2 | 216 | |||
GenBank mRNA | AB025233.1 | - | 2 | 107 | ||
AB025234.1 | - | 3 | 139 | |||
AB025235.1 | - | 3 | 212 | |||
AB025236.1 | - | 3 | 155 | |||
AB025237.1 | - | 3 | 228 | |||
AB025238.1 | - | 2 | 143 | |||
AB025239.1 | - | 2 | 216 | |||
AB025240.1 | - | 3 | 212 | |||
AB025241.1 | - | 3 | 228 | |||
AB025242.1 | - | 2 | 216 | |||
AK026631.1 | - | 3 | 157 | |||
BC014618.2 | - | 2 | 210 | 3531786 | ||
BC040144.1 | - | 2 | 118 | 4390580 | ||
BC051375.1 | - | 2 | 93 | 6651576 | ||
BC065367.1 | - | 2 | 315 | 5814797 | ||
BM927392.1 | - | 2 | 314 | |||
CR407655.1 | - | 1 | 60 | |||
D16581.1 | - | 1 | 86 |