Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002866.4 | NP_002857.1 | 2 | 249 | ||
XM_011528164.1 | XP_011526466.1 | 2 | 246 | |||
GenBank mRNA | AF254795.1 | - | 2 | 209 | ||
AF498931.1 | - | 1 | 80 | |||
AK289559.1 | - | 2 | 227 | |||
AK293467.1 | - | 223 | ||||
BC011782.2 | - | 2 | 214 | 3939176 | ||
DB497194.1 | - | 2 | 259 | |||
M28210.1 | - | 2 | 119 | |||
X13900.1 | - | 2 | 119 |